siRNA Details
| Gene Symbol: Pom121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148932.2 | NP_683734.2 | 11 | 2063 | ||
| GenBank mRNA | AF516680.1 | - | 11 | 2024 | ||
| AK154473.1 | - | 11 | 2064 | |||
| AK172982.1 | - | 10 | 1441 | |||
| BC053101.1 | - | 11 | 2039 | 6405486 | ||

Pom121
Genomic Map