siRNA Details
| Gene Symbol: Olr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301094.1 | NP_001288023.1 | 4 | 417 | ||
| NM_001301096.1 | NP_001288025.1 | 3 | 315 | |||
| NM_138648.2 | NP_619589.2 | 7 | 939 | |||
| GenBank mRNA | AF303744.1 | - | 7 | 928 | ||
| AK154687.1 | - | 7 | 940 | |||
| AK160919.1 | - | 7 | 940 | |||
| HQ603085.1 | - | 3 | 257 | |||
| HQ603086.1 | - | 4 | 359 | |||

Olr1
Genomic Map