siRNA Details
| Gene Symbol: Pip4k2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054051.1 | NP_473392.1 | 10 | 1335 | ||
| GenBank mRNA | AB054987.1 | - | 6 | 728 | ||
| AK089414.1 | - | 10 | 1324 | |||
| AK146559.1 | - | 10 | 1385 | |||
| AK170392.1 | - | 10 | 1263 | |||
| AK171497.1 | - | 10 | 1328 | |||
| AY050219.1 | - | 10 | 1164 | |||
| BC047282.1 | - | 10 | 1335 | 6491155 | ||

Pip4k2b
Genomic Map