siRNA Details
| Gene Symbol: Prkag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170555.1 | NP_001164026.1 | 7 | 607 | ||
| NM_001170556.1 | NP_001164027.1 | 7 | 604 | |||
| NM_145401.2 | NP_663376.2 | 11 | 1686 | |||
| GenBank mRNA | AK032238.1 | - | 9 | 1198 | ||
| AK034765.1 | - | 5 | 272 | |||
| AK154296.1 | - | 11 | 1687 | |||
| AK165834.1 | - | 7 | 546 | |||
| AY348864.1 | - | 7 | 537 | |||
| BC015283.1 | - | 11 | 1474 | 4238045 | ||
| BQ572978.1 | - | 7 | 537 | |||
| BU707674.1 | - | 7 | 507 | |||

Prkag2
Genomic Map