siRNA Details
| Gene Symbol: Prkag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170555.1 | NP_001164026.1 | 10 | 992 | ||
| NM_001170556.1 | NP_001164027.1 | 10 | 989 | |||
| NM_145401.2 | NP_663376.2 | 14 | 2071 | |||
| GenBank mRNA | AK032238.1 | - | 12 | 1583 | ||
| AK034765.1 | - | 8 | 657 | |||
| AK154296.1 | - | 14 | 2072 | |||
| AK165834.1 | - | 10 | 931 | |||
| BC015283.1 | - | 14 | 1859 | 4238045 | ||

Prkag2
Genomic Map