siRNA Details
| Gene Symbol: Prkag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170555.1 | NP_001164026.1 | 10, 11 | 1024 | ||
| NM_001170556.1 | NP_001164027.1 | 10, 11 | 1021 | |||
| NM_145401.2 | NP_663376.2 | 14, 15 | 2103 | |||
| GenBank mRNA | AK032238.1 | - | 12, 13 | 1615 | ||
| AK034765.1 | - | 8, 9 | 689 | |||
| AK154296.1 | - | 14, 15 | 2104 | |||
| AK165834.1 | - | 10, 11 | 963 | |||
| AY348864.1 | - | 10, 11 | 954 | |||
| BC015283.1 | - | 14, 15 | 1891 | 4238045 | ||

Prkag2
Genomic Map