siRNA Details
Gene Symbol: Slco4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148933.1 | NP_683735.1 | 9 | 1969 | ||
GenBank mRNA | AK033598.1 | - | 9 | 1789 | ||
BC030719.1 | - | 9 | 1969 | 4501966 | ||
BC030720.1 | - | 9 | 1918 | 4505508 | ||
BC033602.1 | - | 2 | 171 |
