siRNA Details
| Gene Symbol: Adamts7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003911.2 | NP_001003911.2 | 3 | 718 | ||
| GenBank mRNA | AY551090.1 | - | 3 | 451 | ||
| BC076575.1 | - | 3 | 572 | |||
| BC141173.1 | - | 3 | 494 | 9055785 | ||
| BC145491.1 | - | 3 | 494 | 9054054 | ||

Adamts7
Genomic Map