siRNA Details
| Gene Symbol: Mat2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199274.1 | NP_001186203.1 | 4 | 784 | ||
| NM_134017.2 | NP_598778.1 | 4 | 588 | |||
| GenBank mRNA | AB070267.1 | - | 4 | 555 | ||
| AK075600.1 | - | 4 | 489 | |||
| AK075947.1 | - | 4 | 523 | |||
| AK153126.1 | - | 4 | 544 | |||
| AK162087.1 | - | 4 | 523 | |||
| BC003457.1 | - | 4 | 519 | 2648452 | ||

Mat2b
Genomic Map