siRNA Details
Gene Symbol: Fam210a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153794.4 | NP_722489.1 | 2 | 310 | ||
GenBank mRNA | AK034980.1 | - | 2 | 192 | ||
AK044238.1 | - | 2 | 198 | |||
AK049567.1 | - | 2 | 198 | |||
AK077093.1 | - | 2 | 311 | |||
AK142190.1 | - | 2 | 182 | |||
AK147662.1 | - | 2 | 185 | |||
AK159319.1 | - | 2 | 161 | |||
AK162396.1 | - | 2 | 195 | |||
BC038695.1 | - | 2 | 77 | 1548421 | ||
BC095936.1 | - | 2 | 160 | 3968496 |
