siRNA Details
| Gene Symbol: Rnpepl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181405.4 | NP_852070.3 | 9 | 1795 | ||
| GenBank mRNA | AK075626.1 | - | 5 | 1061 | ||
| AK087430.1 | - | 3 | 298 | |||
| AK154881.1 | - | 6 | 3115 | |||
| BC024079.1 | - | 3 | 308 | |||
| BC031789.1 | - | 9 | 1215 | 4006916 | ||
| CA750931.1 | - | 1 | 162 | |||
| CX218581.1 | - | 5 | 681 | |||

Rnpepl1
Genomic Map