siRNA Details
| Gene Symbol: Dnajc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134081.5 | NP_598842.1 | 3 | 666 | ||
| GenBank mRNA | AK043725.1 | - | 3 | 636 | ||
| AK075979.1 | - | 3 | 627 | |||
| AK077324.1 | - | 3 | 636 | |||
| AK153033.1 | - | 3 | 626 | |||
| AK162075.1 | - | 3 | 541 | |||
| BC014686.1 | - | 3 | 628 | 4481661 | ||
| BC023787.1 | - | 3 | 624 | 5346759 | ||
| BC027012.1 | - | 3 | 619 | 5367851 | ||
| CX238858.1 | - | 3 | 665 | |||

Dnajc9
Genomic Map