siRNA Details
Gene Symbol: Ccdc88a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176841.4 | NP_789811.2 | 29 | 5133 | ||
GenBank mRNA | AB087827.1 | - | 29 | 4957 | ||
AK082771.1 | - | 1 | 177 | |||
BC037020.1 | - | 11 | 1702 |
