siRNA Details
| Gene Symbol: Card11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175362.2 | NP_780571.2 | 14 | 2103 | ||
| GenBank mRNA | AK131134.1 | - | 14 | 2103 | ||
| AY135367.1 | - | 14 | 2014 | |||
| BC096592.1 | - | 14 | 2089 | 5318165 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Card11| Gene Symbol: Card11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175362.2 | NP_780571.2 | 14 | 2103 | ||
| GenBank mRNA | AK131134.1 | - | 14 | 2103 | ||
| AY135367.1 | - | 14 | 2014 | |||
| BC096592.1 | - | 14 | 2089 | 5318165 | ||