siRNA Details
| Gene Symbol: Ston2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175367.6 | NP_780576.1 | 3 | 798 | ||
| GenBank mRNA | AK036612.1 | - | 3 | 799 | ||
| AK160257.1 | - | 3 | 1069 | |||
| BC132531.1 | - | 2 | 384 | 40130808 | ||
| BC138054.1 | - | 2 | 384 | 8861073 | ||

Ston2
Genomic Map