siRNA Details
| Gene Symbol: Ibtk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081282.2 | NP_001074751.1 | 29 | 4560 | ||
| GenBank mRNA | AK031796.1 | - | 18 | 2361 | ||
| AK077387.1 | - | 7 | 703 | |||
| AK129353.1 | - | 26 | 3438 | |||
| BC050255.1 | - | 17 | 3994 | |||
| BC060722.1 | - | 28 | 4354 | |||
| BC137798.1 | - | 29 | 4449 | 8860817 | ||

Ibtk
Genomic Map