siRNA Details
| Gene Symbol: Mtrf1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175374.3 | NP_780583.1 | 2, 3 | 489 | ||
| GenBank mRNA | AK036424.1 | - | 2, 3 | 393 | ||
| AK078897.1 | - | 2, 3 | 457 | |||
| AK185802.1 | - | 2, 3 | 96 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtrf1l| Gene Symbol: Mtrf1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175374.3 | NP_780583.1 | 2, 3 | 489 | ||
| GenBank mRNA | AK036424.1 | - | 2, 3 | 393 | ||
| AK078897.1 | - | 2, 3 | 457 | |||
| AK185802.1 | - | 2, 3 | 96 | |||