siRNA Details
| Gene Symbol: Cdca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110162.1 | NP_001103632.1 | 15 | 2491 | ||
| NM_175384.4 | NP_780593.3 | 15 | 2194 | |||
| GenBank mRNA | AK031313.1 | - | 11 | 1636 | ||
| BC067009.1 | - | 15 | 2332 | |||
| BC075682.1 | - | 15 | 2194 | |||
| BC116298.1 | - | 11 | 1636 | 40095010 | ||
| BC116299.1 | - | 11 | 1636 | 40095011 | ||

Cdca2
Genomic Map