siRNA Details
| Gene Symbol: Zzz3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080755.2 | NP_001074224.1 | 5 | 1989 | ||
| NM_001287139.1 | NP_001274068.1 | 5 | 1931 | |||
| GenBank mRNA | AK034365.1 | - | 2 | 537 | ||
| AK131150.1 | - | 5 | 1927 | |||
| AK132612.1 | - | 2 | 156 | |||
| AK165882.1 | - | 4 | 409 | |||
| BC060614.1 | - | 5 | 1921 | |||

Zzz3
Genomic Map