siRNA Details
| Gene Symbol: Tbc1d10c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178650.3 | NP_848765.2 | 5 | 478 | ||
| GenBank mRNA | AK040487.1 | - | 5 | 479 | ||
| AK075803.1 | - | 5 | 465 | |||
| AK088119.1 | - | 543 | ||||
| AK131177.1 | - | 4 | 459 | |||
| AK146426.1 | - | 5 | 472 | |||
| AK153836.1 | - | 5 | 476 | |||
| AK154444.1 | - | 4 | 456 | |||
| BC094901.1 | - | 4 | 453 | |||

Tbc1d10c
Genomic Map