siRNA Details
| Gene Symbol: Krt75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133357.3 | NP_579935.1 | 9 | 1678 | ||
| GenBank mRNA | AF343088.1 | - | 8 | 1096 | ||
| AK028972.1 | - | 9 | 1679 | |||
| AK037233.1 | - | 9 | 1679 | |||
| AK076367.1 | - | 9 | 1679 | |||
| BC137935.1 | - | 9 | 1679 | 8860954 | ||
| BC137936.1 | - | 9 | 1679 | 8860955 | ||

Krt75
Genomic Map