siRNA Details
| Gene Symbol: Sephs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175400.6 | NP_780609.4 | 3 | 587 | ||
| GenBank mRNA | AK044971.1 | - | 3 | 524 | ||
| AK053638.1 | - | 3 | 622 | |||
| BC065165.1 | - | 3 | 548 | 6417054 | ||
| BC066037.1 | - | 3 | 586 | 6405661 | ||

Sephs1
Genomic Map