siRNA Details
| Gene Symbol: Sephs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175400.6 | NP_780609.4 | 9 | 1482 | ||
| GenBank mRNA | AK044971.1 | - | 9 | 1419 | ||
| AK047631.1 | - | 4 | 513 | |||
| AK053638.1 | - | 9 | 1517 | |||
| AK075655.1 | - | 2 | 184 | |||
| BC065165.1 | - | 9 | 1443 | 6417054 | ||
| BC066037.1 | - | 9 | 1481 | 6405661 | ||

Sephs1
Genomic Map