siRNA Details
| Gene Symbol: Trip11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028446.1 | NP_082722.1 | 11 | 4006 | ||
| GenBank mRNA | BC158098.1 | - | 11 | 4102 | 9088179 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trip11| Gene Symbol: Trip11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028446.1 | NP_082722.1 | 11 | 4006 | ||
| GenBank mRNA | BC158098.1 | - | 11 | 4102 | 9088179 | |