siRNA Details
| Gene Symbol: Trip11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028446.1 | NP_082722.1 | 11 | 1976 | ||
| GenBank mRNA | AK081763.1 | - | 11 | 1889 | ||
| AK139176.1 | - | 1 | 89 | |||
| AK148294.1 | - | 11 | 1952 | |||
| BC158098.1 | - | 11 | 2072 | 9088179 | ||

Trip11
Genomic Map