siRNA Details
| Gene Symbol: Fam118b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286604.1 | NP_001273533.1 | 8 | 1161 | ||
| NM_175411.5 | NP_780620.1 | 5 | 803 | |||
| NM_194257.2 | NP_919233.1 | 6 | 1031 | |||
| GenBank mRNA | AK028018.1 | - | 3 | 552 | ||
| AK079397.1 | - | 8 | 1134 | |||
| AK081206.1 | - | 5 | 779 | |||
| AK153897.1 | - | 6 | 1002 | |||
| AK168168.1 | - | 6 | 1007 | |||
| AK169810.1 | - | 6 | 1006 | |||
| BC051180.1 | - | 6 | 993 | 6315210 | ||

Fam118b
Genomic Map