siRNA Details
| Gene Symbol: Me3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181407.4 | NP_852072.2 | 10 | 1315 | ||
| GenBank mRNA | AK006146.1 | - | 5 | 471 | ||
| AK032196.1 | - | 10 | 1316 | |||
| AK080798.1 | - | 10 | 1248 | |||
| BC099935.1 | - | 10 | 1411 | 30097608 | ||

Me3
Genomic Map