siRNA Details
| Gene Symbol: Mybpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252372.1 | NP_001239301.1 | 9 | 998 | ||
| NM_175418.5 | NP_780627.2 | 10 | 1040 | |||
| GenBank mRNA | AK132188.1 | - | 9 | 872 | ||
| BC062158.1 | - | 10 | 902 | 30134842 | ||

Mybpc1
Genomic Map