siRNA Details
Gene Symbol: Cdcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133974.3 | NP_598735.2 | 9 | 2518 | ||
GenBank mRNA | AK087108.1 | - | 9 | 2419 | ||
BC011340.1 | - | 4 | 1042 | |||
BC039753.1 | - | 3 | 823 | |||
BC050137.1 | - | 2 | 370 | |||
BC085253.1 | - | 9 | 2518 | 5347122 |
