siRNA Details
| Gene Symbol: Txlna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005506.3 | NP_001005506.2 | 2 | 715 | ||
| NM_001199695.1 | NP_001186624.1 | 3 | 432 | |||
| GenBank mRNA | AK078192.1 | - | 3 | 409 | ||
| AK135290.1 | - | 2 | 563 | |||
| AK141252.1 | - | 3 | 366 | |||
| AK150240.1 | - | 3 | 389 | |||
| AK160829.1 | - | 2 | 563 | |||
| AK161649.1 | - | 3 | 395 | |||
| AK168418.1 | - | 2 | 716 | |||
| AK171101.1 | - | 3 | 408 | |||
| BC060227.1 | - | 2 | 550 | 30357541 | ||
| BC065115.1 | - | 2 | 550 | 30362350 | ||

Txlna
Genomic Map