siRNA Details
| Gene Symbol: Ank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034168.1 | NP_001029340.1 | 8 | 2880 | ||
| NM_178655.3 | NP_848770.2 | 23 | 3183 | |||
| GenBank mRNA | AK032060.1 | - | 23 | 3183 | ||
| AK083826.1 | - | 8 | 1439 | |||
| BC043123.1 | - | 8 | 2880 | |||
| BC059251.1 | - | 23 | 3134 | 6405358 | ||

Ank2
Genomic Map