siRNA Details
| Gene Symbol: Arrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177231.2 | NP_796205.1 | 2, 3 | 349 | ||
| NM_178220.3 | NP_835738.1 | 2, 3 | 349 | |||
| GenBank mRNA | AK052588.1 | - | 2, 3 | 126 | ||
| AK090090.1 | - | 2, 3 | 176 | |||
| AK144054.1 | - | 2, 3 | 148 | |||
| AK147508.1 | - | 2, 3 | 350 | |||
| AK147566.1 | - | 2, 3 | 370 | |||
| AK165514.1 | - | 2, 3 | 148 | |||
| BC108969.1 | - | 2, 3 | 116 | 40047224 | ||
| BC108970.1 | - | 2, 3 | 116 | 40047225 | ||

Arrb1
Genomic Map