siRNA Details
| Gene Symbol: Arrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177231.2 | NP_796205.1 | 5, 6 | 644 | ||
| NM_178220.3 | NP_835738.1 | 5, 6 | 644 | |||
| GenBank mRNA | AK052588.1 | - | 5, 6 | 421 | ||
| AK090090.1 | - | 5, 6 | 471 | |||
| AK144054.1 | - | 5, 6 | 443 | |||
| AK147508.1 | - | 5, 6 | 645 | |||
| AK147566.1 | - | 5, 6 | 666 | |||
| AK165514.1 | - | 5, 6 | 443 | |||
| BC108969.1 | - | 5, 6 | 411 | 40047224 | ||
| BC108970.1 | - | 5, 6 | 411 | 40047225 | ||

Arrb1
Genomic Map