siRNA Details
| Gene Symbol: Mzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290452.1 | NP_001277381.1 | 2 | 683 | ||
| NM_001290453.1 | NP_001277382.1 | 2 | 683 | |||
| NM_145819.2 | NP_665818.2 | 2 | 683 | |||
| GenBank mRNA | AB007407.1 | - | 1 | 902 | ||
| AB088478.1 | - | 2 | 584 | |||
| AK030410.1 | - | 2 | 684 | |||
| AK145014.1 | - | 2 | 564 | |||
| AK158837.1 | - | 1 | 1559 | |||
| BC060649.1 | - | 2 | 643 | |||
| BC129904.1 | - | 2 | 433 | 40111654 | ||
| BC129905.1 | - | 2 | 433 | 40111655 | ||

Mzf1
Genomic Map