siRNA Details
| Gene Symbol: Abr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291186.1 | NP_001278115.1 | 7 | 832 | ||
| NM_198018.2 | NP_932135.1 | 11 | 1607 | |||
| NM_198894.2 | NP_942597.1 | 12 | 1493 | |||
| NM_198895.2 | NP_942598.1 | 12 | 1614 | |||
| GenBank mRNA | AK134993.1 | - | 7 | 820 | ||
| AK153615.1 | - | 11 | 2275 | |||
| BC056385.1 | - | 7 | 821 | 6854368 | ||
| BC058708.1 | - | 11 | 1607 | 6839300 | ||
| BC059064.1 | - | 7 | 816 | 6825281 | ||

Abr
Genomic Map