siRNA Details
| Gene Symbol: Phka2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177878.1 | NP_001171349.1 | 6 | 972 | ||
| NM_001177879.1 | NP_001171350.1 | 7 | 786 | |||
| NM_172783.3 | NP_766371.1 | 6 | 972 | |||
| GenBank mRNA | AK052346.1 | - | 6 | 717 | ||
| AK052992.1 | - | 6 | 973 | |||
| AK165506.1 | - | 7 | 787 | |||
| BC050040.1 | - | 7 | 745 | 6431743 | ||
| BC051167.1 | - | 1 | 212 | |||
| BC058254.1 | - | 6 | 941 | |||

Phka2
Genomic Map