siRNA Details
| Gene Symbol: Phka2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177878.1 | NP_001171349.1 | 18 | 2207 | ||
| NM_001177879.1 | NP_001171350.1 | 19 | 2021 | |||
| NM_172783.3 | NP_766371.1 | 18 | 2207 | |||
| GenBank mRNA | AK052346.1 | - | 18 | 1952 | ||
| AK052992.1 | - | 18 | 2208 | |||
| AK165506.1 | - | 19 | 2022 | |||
| BC050040.1 | - | 19 | 1980 | 6431743 | ||
| BC051167.1 | - | 13 | 1449 | |||

Phka2
Genomic Map