siRNA Details
| Gene Symbol: Pygl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133198.2 | NP_573461.2 | 10, 11 | 1347 | ||
| GenBank mRNA | AF288783.1 | - | 10, 11 | 1289 | ||
| AK140321.1 | - | 10, 11 | 1294 | |||
| AK145989.1 | - | 10, 11 | 1348 | |||
| AK149491.1 | - | 1522 | ||||
| AK167337.1 | - | 10, 11 | 1294 | |||
| BC013636.1 | - | 10, 11 | 1281 | 4196237 | ||

Pygl
Genomic Map