siRNA Details
| Gene Symbol: Pygl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133198.2 | NP_573461.2 | 1 | 173 | ||
| GenBank mRNA | AF288783.1 | - | 1 | 115 | ||
| AK041189.1 | - | 1 | 120 | |||
| AK140321.1 | - | 1 | 120 | |||
| AK145989.1 | - | 1 | 174 | |||
| AK167337.1 | - | 1 | 120 | |||
| BC013636.1 | - | 1 | 107 | 4196237 | ||

Pygl
Genomic Map