siRNA Details
Gene Symbol: Gpr18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182806.1 | NP_877958.1 | 2 | 653 | ||
GenBank mRNA | AK156039.1 | - | 2 | 726 | ||
AK156868.1 | - | 2 | 718 | |||
AK157029.1 | - | 2 | 661 | |||
AK172401.1 | - | 2 | 656 | |||
BC034872.1 | - | 2 | 653 | 1397739 |
