siRNA Details
| Gene Symbol: Slc35b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016752.1 | NP_058032.1 | 3 | 408 | ||
| GenBank mRNA | AK002961.1 | - | 3 | 402 | ||
| AK007711.1 | - | 3 | 414 | |||
| AK160124.1 | - | 3 | 474 | |||
| AK183623.1 | - | 2 | 192 | |||
| AK184790.1 | - | 2 | 192 | |||
| AK185566.1 | - | 2 | 189 | |||
| BC002029.1 | - | 3 | 389 | 3592141 | ||
| D87990.1 | - | 3 | 408 | |||

Slc35b1
Genomic Map