siRNA Details
| Gene Symbol: Msra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253712.1 | NP_001240641.1 | 1 | 267 | ||
| NM_001253714.1 | NP_001240643.1 | 1 | 253 | |||
| NM_001253715.1 | NP_001240644.1 | 1 | 253 | |||
| NM_026322.4 | NP_080598.2 | 6 | 666 | |||
| GenBank mRNA | AK002356.1 | - | 1 | 142 | ||
| AK009822.1 | - | 1 | 144 | |||
| AK030278.1 | - | 1 | 145 | |||
| AK049714.1 | - | 1 | 268 | |||
| BB652761.1 | - | 1 | 265 | |||
| BC014738.1 | - | 1 | 212 | 4239349 | ||
| BC089311.1 | - | 6 | 666 | 6412625 | ||
| CB953198.1 | - | 1 | 124 | |||

Msra
Genomic Map