siRNA Details
| Gene Symbol: Bcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081412.2 | NP_001074881.1 | 16 | 3043 | ||
| GenBank mRNA | AK129482.1 | - | 13 | 1233 | ||
| BC048842.1 | - | 4 | 287 | |||
| BC060270.1 | - | 16 | 2281 | |||
| BC131682.1 | - | 17 | 2448 | 40111128 | ||

Bcr
Genomic Map