siRNA Details
| Gene Symbol: Acat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144784.3 | NP_659033.1 | 6 | 620 | ||
| GenBank mRNA | AJ510150.1 | - | 6 | 612 | ||
| AK032097.1 | - | 6 | 591 | |||
| AK075593.1 | - | 6 | 593 | |||
| AK081715.1 | - | 6 | 620 | |||
| AK146241.1 | - | 6 | 575 | |||
| AK163402.1 | - | 6 | 624 | |||
| AK169771.1 | - | 6 | 613 | |||
| BC024429.1 | - | 6 | 623 | |||
| BC024763.1 | - | 6 | 592 | 5365469 | ||
| BC029345.1 | - | 3 | 281 | |||

Acat1
Genomic Map