siRNA Details
| Gene Symbol: Acat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144784.3 | NP_659033.1 | 5 | 428 | ||
| GenBank mRNA | AJ510150.1 | - | 5 | 420 | ||
| AK032097.1 | - | 5 | 399 | |||
| AK075593.1 | - | 5 | 400 | |||
| AK081715.1 | - | 5 | 428 | |||
| AK146241.1 | - | 5 | 383 | |||
| AK163402.1 | - | 5 | 432 | |||
| AK169771.1 | - | 5 | 421 | |||
| BC024429.1 | - | 5 | 431 | |||
| BC024763.1 | - | 5 | 400 | 5365469 | ||
| BC029345.1 | - | 2 | 89 | |||

Acat1
Genomic Map