siRNA Details
| Gene Symbol: Hivep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007772.2 | NP_031798.2 | 4 | 532 | ||
| GenBank mRNA | AK039808.1 | - | 4 | 532 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hivep1| Gene Symbol: Hivep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007772.2 | NP_031798.2 | 4 | 532 | ||
| GenBank mRNA | AK039808.1 | - | 4 | 532 | ||