siRNA Details
| Gene Symbol: Adarb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024837.2 | NP_001020008.1 | 3, 4 | 432 | ||
| NM_130895.3 | NP_570965.2 | 3, 4 | 432 | |||
| GenBank mRNA | AF291049.1 | - | 2, 3 | 267 | ||
| AF403106.2 | - | 3, 4 | 384 | |||
| AF403107.2 | - | 3, 4 | 384 | |||
| AK138318.1 | - | 3, 4 | 425 | |||
| AK141777.1 | - | 3, 4 | 404 | |||
| AK147298.1 | - | 4, 5 | 511 | |||
| FJ169505.1 | - | 2, 3 | 207 | |||

Adarb1
Genomic Map