siRNA Details
| Gene Symbol: Aldh7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127338.1 | NP_001120810.1 | 13 | 1208 | ||
| NM_138600.4 | NP_613066.2 | 11 | 1014 | |||
| GenBank mRNA | AK004991.1 | - | 13 | 1142 | ||
| AK145873.1 | - | 11 | 957 | |||
| AK169195.1 | - | 11 | 957 | |||
| AK183099.1 | - | 1 | 15 | |||

Aldh7a1
Genomic Map