siRNA Details
| Gene Symbol: Adgrv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054053.4 | NP_473394.3 | 24 | 5361 | ||
| GenBank mRNA | AF405692.1 | - | 20 | 5506 | ||
| AF405693.1 | - | 14 | 3160 | |||
| AF435926.1 | - | 24 | 5362 | |||
| JQ013292.1 | - | 1 | 22 | |||

Adgrv1
Genomic Map