siRNA Details
| Gene Symbol: Pcca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144844.2 | NP_659093.2 | 3, 4 | 252 | ||
| GenBank mRNA | AK148002.1 | - | 3, 4 | 242 | ||
| AK212580.1 | - | 3, 4 | 156 | |||
| AY046947.1 | - | 3, 4 | 269 | |||
| BC006915.1 | - | 3, 4 | 249 | 3601148 | ||
| BC049802.1 | - | 3, 4 | 218 | 6332730 | ||
| BY263345.1 | - | 3, 4 | 252 | |||

Pcca
Genomic Map